Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029045.5 | NP_001024216.1 | 5 - 6 | 701 | 84 | |
NM_001269959.3 | NP_001256888.1 | 2 - 3 | 238 | 84 | ||
NM_075449.9 | NP_507850.2 | 2 - 3 | 240 | 84 | ||
GenBank mRNA | AF031834.1 | - | 2 - 3 | 253 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |