Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029045.5 | NP_001024216.1 | 7 - 8 | 1457 | 75 | |
NM_001269959.3 | NP_001256888.1 | 4 - 5 | 994 | 75 | ||
NM_075449.9 | NP_507850.2 | 4 - 5 | 996 | 75 | ||
GenBank mRNA | AF031834.1 | - | 4 - 5 | 1009 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |