Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029041.3 | NP_001024212.1 | 2 - 3 | 128 | 80 | |
NM_001029042.5 | NP_001024213.1 | 2 - 3 | 126 | 80 | ||
NM_001330887.2 | NP_001317838.1 | 1 - 2 | 90 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |