Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029041.3 | NP_001024212.1 | 1 - 2 | 55 | 67 | |
NM_001029042.5 | NP_001024213.1 | 1 - 2 | 53 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |