Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270018.2 | NP_001256947.1 | 10 - 11 | 1336 | 62 | |
NM_001270019.3 | NP_001256948.1 | 3 - 4 | 338 | 62 | ||
NM_001270020.4 | NP_001256949.1 | 2 - 3 | 288 | 62 | ||
NM_001373209.3 | NP_001360578.1 | 11 | 1343 | 62 | ||
GenBank mRNA | AF293972.1 | - | 10 - 11 | 1356 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |