Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pha-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047649.4 | NP_001041114.1 | 5 - 6 | 895 | 64 | |
NM_001047650.4 | NP_001041115.1 | 4 - 5 | 703 | 64 | ||
NM_001047651.6 | NP_001041116.1 | 3 - 4 | 613 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |