Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277022.3 | NP_001263951.1 | 13 - 14 | 2514 | 109 | |
NM_001277023.4 | NP_001263952.1 | 13 - 14 | 2526 | 109 | ||
NM_075617.5 | NP_508018.2 | 13 - 14 | 2572 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |