Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: num-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136432.5 | NP_001129904.1 | 3 - 4 | 975 | 70 | |
NM_075620.6 | NP_508021.1 | 1 - 2 | 151 | 70 | ||
GenBank mRNA | AF286205.1 | - | 1 - 2 | 151 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |