Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrde-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075691.7 | NP_508092.1 | 6 - 7 | 1889 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |