Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075777.7 | NP_508178.2 | 11 - 12 | 1325 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |