Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373217.4 | NP_001360743.1 | 1 - 2 | 97 | 114 | |
NM_001380899.1 | NP_001367822.1 | 1 - 2 | 99 | 114 | ||
NM_075777.7 | NP_508178.2 | 1 - 2 | 51 | 114 | ||
NM_075778.5 | NP_508179.2 | 1 - 2 | 104 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |