Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380905.1 | NP_001367433.1 | 6 - 7 | 760 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |