Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrt-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075897.5 | NP_508298.2 | 8 - 9 | 867 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |