Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383530.1 | 3 - 4 | 572 | 81 | ||
GenBank mRNA | AJ427855.1 | - | 3 - 4 | 574 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |