Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029525.4 | NP_001024696.1 | 7 - 8 | 930 | 135 | |
NM_001029526.4 | NP_001024697.1 | 6 - 7 | 762 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |