Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374981.3 | NP_001362097.1 | 13 - 14 | 3856 | 83 | |
NM_076103.6 | NP_508504.2 | 14 - 15 | 3878 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |