Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076125.4 | NP_508526.2 | 10 - 11 | 1450 | 117 | |
GenBank mRNA | AF083645.1 | - | 10 - 11 | 1442 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |