Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029758.3 | NP_001024929.1 | 2 - 3 | 387 | 135 | |
NM_001029759.5 | NP_001024930.1 | 2 - 3 | 387 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |