Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029508.6 | NP_001024679.1 | 7 - 8 | 1221 | 98 | |
NM_001383562.1 | 7 - 8 | 1217 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |