Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsp-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380953.1 | NP_001367834.1 | 1 - 2 | 121 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |