Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375027.2 | NP_001362033.1 | 13 - 14 | 5364 | 98 | |
NM_001375029.2 | NP_001362032.1 | 12 - 13 | 5157 | 98 | ||
NM_001375030.2 | NP_001362152.1 | 12 - 13 | 5315 | 98 | ||
NM_001375031.2 | NP_001362153.1 | 11 - 12 | 5065 | 98 | ||
NM_001383570.1 | 13 - 14 | 5668 | 98 | |||
NM_076447.6 | NP_508848.2 | 14 - 15 | 5702 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |