Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375027.2 | NP_001362033.1 | 4 - 5 | 556 | 64 | |
NM_001375029.2 | NP_001362032.1 | 3 - 4 | 349 | 64 | ||
NM_001375030.2 | NP_001362152.1 | 3 - 4 | 507 | 64 | ||
NM_001375031.2 | NP_001362153.1 | 2 - 3 | 257 | 64 | ||
NM_001383570.1 | 4 - 5 | 860 | 64 | |||
NM_076447.6 | NP_508848.2 | 5 - 6 | 894 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |