Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076449.6 | NP_508850.2 | 9 - 10 | 1271 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |