Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076469.6 | NP_508870.3 | 8 - 9 | 1228 | 63 | |
NM_076470.7 | NP_508871.1 | 8 - 9 | 1230 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |