Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ift-81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076499.9 | NP_508900.6 | 16 - 17 | 1659 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |