Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sto-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270091.4 | NP_001257020.1 | 7 - 8 | 812 | 102 | |
NM_001270092.5 | NP_001257021.1 | 7 - 8 | 2523 | 102 | ||
NM_001270093.4 | NP_001257022.1 | 7 - 8 | 805 | 102 | ||
NM_001270094.2 | NP_001257023.1 | 6 - 7 | 696 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |