Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029747.6 | NP_001024918.1 | 10 - 11 | 1395 | 150 | |
NM_001307913.5 | NP_001294842.1 | 7 - 8 | 1646 | 150 | ||
NM_001330899.4 | NP_001317909.1 | 8 - 9 | 1222 | 150 | ||
NM_001330900.4 | NP_001317910.1 | 4 - 5 | 847 | 150 | ||
NM_001380981.1 | NP_001368074.1 | 8 - 9 | 1174 | 150 | ||
NM_001383586.1 | 9 - 10 | 1276 | 150 | |||
NR_138511.1 | 9 - 10 | 1447 | 150 | |||
NR_138512.1 | 9 - 10 | 1611 | 150 | |||
GenBank mRNA | AF097319.1 | - | 9 - 10 | 1326 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |