Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076593.8 | NP_508994.1 | 2 - 3 | 223 | 93 | |
GenBank mRNA | AB126261.1 | - | 2 - 3 | 203 | 93 |
Gene Symbol: acs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076592.9 | NP_508993.2 | 2 - 3 | 218 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |