Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373314.3 | NP_001360738.1 | 13 | 1309 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Amplicon spans exons and probe does not span exons |