Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ets-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373331.1 | NP_001359862.1 | 9 - 10 | 1012 | 71 | |
NM_076698.6 | NP_509099.2 | 6 - 7 | 602 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |