Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076728.4 | NP_509129.1 | 4 - 5 | 531 | 95 | |
GenBank mRNA | Y08637.1 | - | 531 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |