Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029655.6 | NP_001024826.2 | 5 - 6 | 923 | 93 | |
NM_001129697.5 | NP_001123169.1 | 5 - 6 | 923 | 93 | ||
NM_001129698.4 | NP_001123170.1 | 5 - 6 | 881 | 93 | ||
NM_001373337.3 | NP_001360088.1 | 6 | 929 | 93 | ||
GenBank mRNA | AY887906.1 | - | 5 - 6 | 945 | 93 | |
AY954522.1 | - | 5 - 6 | 945 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |