Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bar-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076805.8 | NP_509206.1 | 7 - 8 | 798 | 154 | |
GenBank mRNA | AF063646.1 | - | 7 - 8 | 823 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |