Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076828.8 | NP_509229.1 | 8 - 9 | 1337 | 92 | |
GenBank mRNA | AY091467.1 | - | 7 - 8 | 1268 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |