Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029600.6 | NP_001024771.1 | 3 - 4 | 765 | 70 | |
NR_132545.1 | 2 - 3 | 420 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |