Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076966.3 | NP_509367.1 | 2 - 3 | 197 | 59 | |
GenBank mRNA | AF037063.1 | - | 1 - 2 | 102 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |