Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270133.3 | NP_001257062.1 | 2 - 3 | 169 | 65 | |
NM_001270134.3 | NP_001257063.1 | 1 - 2 | 77 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |