Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029659.5 | NP_001024830.1 | 10 - 11 | 1694 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |