Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vav-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047757.3 | NP_001041222.1 | 15 - 16 | 2694 | 125 | |
NM_001373412.3 | NP_001359918.1 | 16 - 17 | 2850 | 125 | ||
NM_001373413.3 | NP_001360680.1 | 16 - 17 | 2837 | 125 | ||
GenBank mRNA | DQ136172.1 | - | 16 - 17 | 2788 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |