Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083286.4 | NP_001076755.1 | 2 - 3 | 449 | 92 | |
NM_001083287.6 | NP_001076756.1 | 1 - 2 | 79 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |