Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129721.4 | NP_001123193.2 | 12 - 13 | 1586 | 95 | |
NM_001313458.3 | NP_001300387.1 | 13 - 14 | 1628 | 95 | ||
NM_001313459.3 | NP_001300388.1 | 9 - 10 | 1361 | 95 | ||
NM_077273.4 | NP_509674.2 | 13 - 14 | 1845 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |