Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077293.8 | NP_509694.1 | 3 - 4 | 491 | 114 | |
GenBank mRNA | AF042389.1 | - | 3 - 4 | 445 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |