Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erd-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381076.2 | NP_001366716.1 | 6 - 7 | 1538 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |