Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: git-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077360.8 | NP_509761.2 | 5 - 6 | 473 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |