Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbo-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077429.8 | NP_509830.2 | 10 - 11 | 1679 | 86 | |
GenBank mRNA | AF497831.1 | - | 10 - 11 | 1701 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |