Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbo-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077429.8 | NP_509830.2 | 12 - 13 | 2031 | 63 | |
GenBank mRNA | AF497831.1 | - | 12 - 13 | 2053 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |