Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077448.6 | NP_509849.1 | 6 - 7 | 1692 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |