Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apt-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077452.6 | NP_509853.2 | 10 - 11 | 1453 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |