Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oct-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029821.6 | NP_001024992.2 | 1 - 2 | 133 | 66 | |
NM_077498.9 | NP_509899.3 | 6 - 7 | 984 | 66 | ||
GenBank mRNA | AF199013.1 | - | 1 - 2 | 133 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |