Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oct-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029821.6 | NP_001024992.2 | 4 - 5 | 762 | 96 | |
NM_077498.9 | NP_509899.3 | 9 - 10 | 1613 | 96 | ||
GenBank mRNA | AF199013.1 | - | 4 - 5 | 762 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |